Coulomat oils for Karl Fischer titration, HYDRANAL™
Lieferant: Honeywell Chemicals
34868-500ML 34868-100ML 34868-6X100ML 34868-6X500ML
34868-500MLEA 131.07 EUR
34868-500ML 34868-100ML 34868-6X100ML 34868-6X500ML
Coulomat oils for Karl Fischer titration, HYDRANAL™
Karl Fischer Titriermittel und Reagenzien
Coulomat oil is a reagent for coulometric KF titration in oils (anolyte solution), preferred to use for the cells with diaphragm.