Natriumbenzoat 99,0-101,0% (durch Trockensubstanz) NF, FCC, J.T.Baker®

Lieferant: Avantor
Achtung

3500-05EA 377.94 EUR
3500-05
Natriumbenzoat 99,0-101,0% (durch Trockensubstanz) NF, FCC, J.T.Baker®
Natriumbenzoat
Formel: C₆H₅COONa
Schmelzpunkt: 410…430 °C
Dichte: 1,44 g/cm³ (20 °C)
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012463
CAS-Nummer: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Bestellen

Spezifikation Testergebnisse

Meets N.F. Requirements
GMP Manufactured Product
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Identification C Passes Test
NF - Alkalinity Passes Test
NF - Water (H₂O) ≤1.5 %
NF - Assay (C₆H₅COONa) (anhydrous basis) 99.0 - 101.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Identification C Passes Test
FCC - Alkalinity ≤0.04 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Water (H₂O) ≤1.5 %
FCC - Assay (C₆H₅COONa) (anhydrous basis) 99.0 - 101.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR