Natriumacetat 0.1 M CH3COONa in Essigsäure volumetrische Lösung, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Essigsäure Natriumsalz
35208-1L
35208-1LEA
149.29
EUR
35208-1L
Natriumacetat 0.1 M CH3COONa in Essigsäure volumetrische Lösung, Fluka™
Natriumacetat
Honeywell's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Beilstein Number: 3786729
Formel:
H₃CCOONa Molecular Weight: 82.03 g/mol Siedepunkt: 117…118 °C Schmelzpunkt: 16.2 °C Dichte: 1.05 g/cm³ Flammpunkt: 19.9 %(V) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137248 CAS-Nummer: 127-09-3 EINECS: 200-580-7 UN: 2789 ADR: 8 II Merck Index: 12,08711 |
Spezifikation Testergebnisse
Factor | 0.99 - 1.01 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...