Natriumhydroxid 1 M NaOH (1N) aqueous solution Reag. Ph. Eur. Volumetric, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Caustic soda

35256-5L 35256-10L-VP 35256-1L 35256-4X5L 35256-6X1L 35256-5L-VP
35256-5LEA 120.14 EUR
35256-5L 35256-10L-VP 35256-1L 35256-4X5L 35256-6X1L 35256-5L-VP
Natriumhydroxid 1 M NaOH (1N) aqueous solution Reag. Ph. Eur. Volumetric, Fluka™
Sodium hydroxide
Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 100 °C
Schmelzpunkt: 0 °C
Dichte: 1.04 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

Vapor Pressure 3 mmHg ( 37 °C)
Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR