Natriumhydrogensulfat Monohydrat 35,0 - 36,5% (durch Acidimetrie), Kristalle, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor
Gefahr

3534-01EA 202.9 EUR
3534-01
Natriumhydrogensulfat Monohydrat 35,0 - 36,5% (durch Acidimetrie), Kristalle, BAKER ANALYZED®, J.T.Baker®
Natriumhydrogensulfat Monohydrat

Crystals.

Formel: NaHSO₄·H₂O
Schmelzpunkt: 183 °C
Dichte: 1,8 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149210
CAS-Nummer: 10034-88-5
EINECS: 231-665-7
UN: 3260
ADR: 8,III
Merck Index: 12,08729

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 35.0 - 36.5 %
Assay (H₂SO₄) (by acidimetry) 35.0 - 36.5 %
Insoluble Matter and NH₄OH Precipitate ≤0.005 %
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Calcium and Magnesium (as Ca) (by EDTA titration) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - Arsenic (As) ≤1 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR