Soap solution according to boutron and boudet-Lösung DIN 8106, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

35366-1L
35366-1LEA 81.38 EUR
35366-1L
Soap solution according to boutron and boudet-Lösung DIN 8106, Fluka™
Soap solution according to boutron and boudet
Siedepunkt: 78 °C (1,013 hPa)
Dichte: 0,93 g/cm3 (20 °C)
Flammpunkt: 22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00148418
UN: 1170
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Factor 0.99 - 1.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR