Organic reference standard, Citrinin solution, analytical standard, Supelco®
Lieferant: Merck
35410-1ML
35410-1MLEA 544 EUR
35410-1ML
Organic reference standard, Citrinin solution, analytical standard, Supelco®
Standards Organische Standards
Citrinin is a hepato-nephrotoxic mycotoxin. It can be produced by several filamentous fungi genera of Monauscus, Aspergillus, and Penicillium, which is commonly found as a contaminant in stored grains, foods, feedstuffs and also in biological fluids.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=327b6e045f7655ecf19dbf9924af19aaa67dbceb&vid=327b6e045f7655ecf19dbf9924af19aaa67dbceb&prid=web&clid=SAPDAM