Kupfer(II)acetat Monohydrat ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cupriaacetat Monohydrat, Kupferdiacetat Monohydrat, Essigsäure Kupfer(II)salz Monohydrat, Grünspan
35481.06
35481.14
AB207600.0005
35481.06EA
93.8
EUR
35481.06
35481.14
ABCRAB207600.0005
Kupfer(II)acetat Monohydrat ≥99,9% (Metall-Basis)
Kupfer(II)acetat Monohydrat
Purity: 99.9%
CAS No.: 6046-93-1
Molecular Formula: C4H6CuO4.H2O
Formula Weight: 199.65 (181.64anhy)
Physical Form: -4 Mesh
Density: 1.882
Appearance: Blue-green
Odour: Odourless
pH: 5.2-5.5
Melting Point: 115°
Merck Reference: 14,2624
UN No.: 3077
MDL No.: MFCD00149570
Formel:
(H₃CCOO)₂Cu·H₂O Schmelzpunkt: 115 °C Dichte: 1,882 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149570 CAS-Nummer: 6046-93-1 EINECS: 205-553-3 UN: 3077 ADR: 9,III Merck Index: 14,2624 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...