Natriummetabisulfit NF, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Natriumpyrosulfit, Pyrosulfit, Natriumdisulfit, di-Natriumdisulfit, Disulfit

3550-05EA 207.3 EUR
3550-05
Natriummetabisulfit NF, J.T.Baker®
Natriummetabisulfit
Formel: Na₂S₂O₅
Molecular Weight: 190,11 g/mol
Dichte: 1,48 g/cm³
MDL: MFCD00167602
CAS-Nummer: 7681-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Limit of Chloride ≤0.05 %
NF - Iron (Fe) ≤0.002 %
NF - Limit of Thiosulfate ≤0.05 %
NF - Assay (SO₂) 65.0 - 67.4 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR