Natriummetabisulfit NF, Multikompendial, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Natriumpyrosulfit, Pyrosulfit, Natriumdisulfit, di-Natriumdisulfit, Disulfit

Veränderungsmanagement-Kategorie= R

3551-05EA 152.08 EUR
3551-05
Natriummetabisulfit NF, Multikompendial, J.T.Baker®
Natriummetabisulfit

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets N.F. Requirements

Formel: Na₂S₂O₅
Molecular Weight: 190,11 g/mol
Schmelzpunkt: 150 °C
Dichte: 1,48 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00167602
CAS-Nummer: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product Passes Test
Meets J.P. Chemical Specifications Passes Test
Meets B.P. Chemical Specifications = 0.05 %
Meets E.P. Chemical Specifications = 0.002 %
Meets N.F. Requirements = 0.05 %
CAUTION: For Manufacturing, processing or repackaging = 0.002 %
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Limit of Chloride ≤ 0.05 %
NF - Iron (Fe) ≤ 0.002 %
NF - Limit of Thiosulfate ≤ 0.05 %
NF - Assay (SO₂) 65.0 - 67.4 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Thiosulfate Passes Test
EP/BP - Iron (Fe) ≤ 20 ppm
EP/BP - Assay (Na₂S₂O₅) 95.0 - 100.5 %
EP/BP - pH 3.5 - 5.0
JP - Assay (Na₂S₂O₅) 95.0 - 100.5 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Thiosulfate Passes Test
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Iron (Fe) ≤ 20 ppm
JP - Arsenic (As) = 4 ppm
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. = 20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR