Natriummetabisulfit ≥97,0%, körnig, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Natriumpyrosulfit, Pyrosulfit, Natriumdisulfit, di-Natriumdisulfit, Disulfit

3552-07EA 0 EUR
3552-07
Natriummetabisulfit ≥97,0%, körnig, BAKER ANALYZED® ACS, J.T.Baker®
Natriummetabisulfit

Granular. Lot analysis on label.

Formel: Na₂S₂O₅
Molecular Weight: 190,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00167602
CAS-Nummer: 7681-57-4
EINECS: 231-673-0
Merck Index: 13,08712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use ≥ 97.0 %
Assay (Na₂S₂O₅) ≥97.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.05 %
Thiosulfate (S₂O₃) ≤0.05 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.002 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR