Sulphate, Ion chromatography standard solution, Specpure®, SO₄⁻² 1000 µg/ml
Lieferant: Thermo Fisher Scientific
35547.AE
35547.AP
35547.AEEA
142
EUR
35547.AE
35547.AP
Sulphate, Ion chromatography standard solution, Specpure®, SO₄⁻² 1000 µg/ml
Standards
Anionische Standards
Sulfat-Standards
Molecular Formula: K₂SO₄ in H₂O
Physical Form: Liquid
MDL No.: MFCD00145343
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...