Nitrate, Ion chromatography standard solution, Specpure®, NO₃- 1000 µg/ml
Lieferant: Thermo Fisher Scientific
35549.AE
35549.AP
35549.AEEA
139
EUR
35549.AE
35549.AP
Nitrate, Ion chromatography standard solution, Specpure®, NO₃- 1000 µg/ml
Standards
Anionische Standards
Nitrat-Standards
Molecular Formula: NaNO₃ in H₂O
Physical Form: Liquid
MDL No.: MFCD00144920
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...