Aluminiumchlorid solution_standardised 6.0 N aqueous solution

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Salzsäure Aluminiumsalz

35568.AE 35568.AK
35568.AEEA 71.5 EUR
35568.AE 35568.AK
Aluminiumchlorid solution_standardised 6.0 N aqueous solution
Aluminiumchlorid

Molecular Formula: AlCl3 Formula Weight: 133.34 Physical Form: Liquid Odour: Odourless UN No.: 2581 MDL No.: MFCD00003422

Formel: AlCl₃
Molecular Weight: 133.34 g/mol
Siedepunkt: 180
Lagertemperatur: Raumtemperatur
MDL: MFCD00003422
CAS-Nummer: 7446-70-0
EINECS: 231-208-1
UN: 2581
ADR: 8 III
Merck Index: 13,00335

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR