Ammoniaklösung 50% (v/v) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ammoniumhydroxid, Ammoniak
35574.AP
35574.K2
35574.K7
35574.APEA
62.3
EUR
35574.AP
35574.K2
35574.K7
Ammoniaklösung 50% (v/v) in wässriger Lösung
Ammoniaklösung
CAS No.: 1336-21-6
Molecular Formula: H5NO
Formula Weight: 35.05
Physical Form: Liquid
Density: 0.94
Appearance: Colourless
UN No.: UN2672
MDL No.: MFCD00066650
Formel:
NH₃ Dichte: 0,94 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066650 CAS-Nummer: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...