Ammoniumdimolybdat 10% w/v aqueous solution

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ammoniummolybdat (di), di-Ammoniumdimolybdat

35579.AP 35579.K2
35579.APEA 70.9 EUR
35579.AP 35579.K2
Ammoniumdimolybdat 10% w/v aqueous solution
Ammonium dimolybdate

Molecular Formula: (NH4)2Mo2O7 Formula Weight: 339.95 Physical Form: Liquid MDL No.: MFCD00010882

Formel: H₈Mo₂N₂O₇
Molecular Weight: 339.95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010882
CAS-Nummer: 27546-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR