Natriumhydroxid solution_standardised 1.0 N aqueous solution

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Caustic soda

35629.K2
35629.K2EA 31.8 EUR
35629.K2
Natriumhydroxid solution_standardised 1.0 N aqueous solution
Sodium hydroxide

CAS No.: 1310-73-2 Molecular Formula: HNaO Formula Weight: 40.00 Physical Form: Liquid Appearance: Colourless UN No.: 3266 MDL No.: MFCD00003548

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390
Schmelzpunkt: 323
Dichte: 1.04 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 3266
ADR: 8 III
Merck Index: 14,08627

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR