Natriumhydroxid solution_standardised 5.0 N aqueous solution
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Caustic soda
35633.K2
35633.K2EA
44.3
EUR
35633.K2
Natriumhydroxid solution_standardised 5.0 N aqueous solution
Sodium hydroxide
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Liquid
UN No.: 1824
MDL No.: MFCD00003548
Formel:
NaOH Molecular Weight: 40 g/mol Siedepunkt: 1390 Schmelzpunkt: 323 Dichte: ≤1.11 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003548 CAS-Nummer: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8 II Merck Index: 14,08627 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...