Natriumthiosulfat solution_standardised 1.0 N aqueous solution

Lieferant: Thermo Fisher Scientific

Synonyme: di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit

35647.K2
35647.K2EA 44.7 EUR
35647.K2
Natriumthiosulfat solution_standardised 1.0 N aqueous solution
Natriumthiosulfat

CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00003499

Formel: Na₂S₂O₃
Molecular Weight: 158.11 g/mol
Siedepunkt: 100
Schmelzpunkt: 48
Lagertemperatur: Raumtemperatur
MDL: MFCD00003499
CAS-Nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR