Hexaborsilicid ≥98% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

35670.09 35670.18
35670.09EA 88.1 EUR
35670.09 35670.18
Hexaborsilicid ≥98% (Metall-Basis)
Hexaboron silicide

Purity: 98% CAS No.: 12008-29-6 Molecular Formula: B6Si, may contain traces of Si, other B-Si phases Formula Weight: 92.95 Physical Form: -200 Mesh Powder Odour: Odourless MDL No.: MFCD00180907

Formel: B₆Si
Lagertemperatur: Raumtemperatur
MDL: MFCD00180907
CAS-Nummer: 12008-29-6
EINECS: 234-535-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR