Bleititanat 99,9 Metals basis
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Blei(II)titanat(IV), Blei(II)titan(IV)oxid, Blei(II)orthotitanat, Blei(II)titanoxid, Bleititantrioxid, Blei(II)titanat
35671.18
35671.30
35671.18EA
91.1
EUR
35671.18
35671.30
Bleititanat 99,9 Metals basis
Bleititanat
Purity: 99.9%
CAS No.: 12060-00-3
Molecular Formula: PbTiO3
Formula Weight: 303.09
Physical Form: -200 Mesh Powder
Density: 7.520
Appearance: Yellow
Odour: Odourless
UN No.: 3288
MDL No.: MFCD00011159
Formel:
O₃PbTi Molecular Weight: 303.08 g/mol Schmelzpunkt: 1120 Dichte: 7.520 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011159 CAS-Nummer: 12060-00-3 EINECS: 235-038-9 UN: 3288 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...