di-Natrium-β-glycerophosphat Pentahydrat, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: ortho-Phosphorsäure(2-hydroxy-1-(hydroxymethyl)ethyl)ester Dinatriumsalz Pentahydrat

35675-100 35675-250 35675-50 35675.50G
35675-100EA 88.5 EUR
35675-100 35675-250 35675-50 35675.50G
di-Natrium-β-glycerophosphat Pentahydrat, Millipore®
di-Natrium-β-glycerophosphat Pentahydrat
Formel: C₃H₇Na₂O₆P·5H₂O
Molecular Weight: 306.11 g/mol
MDL: MFCD00002135
CAS-Nummer: 13408-09-8
EINECS: 212-464-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR