di-Natrium-β-glycerophosphat Pentahydrat, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
ortho-Phosphorsäure(2-hydroxy-1-(hydroxymethyl)ethyl)ester Dinatriumsalz Pentahydrat
35675-100
35675-250
35675-50
35675.50G
35675-100EA
88.5
EUR
35675-100
35675-250
35675-50
35675.50G
di-Natrium-β-glycerophosphat Pentahydrat, Millipore®
di-Natrium-β-glycerophosphat Pentahydrat
Formel:
C₃H₇Na₂O₆P·5H₂O Molecular Weight: 306.11 g/mol |
MDL:
MFCD00002135 CAS-Nummer: 13408-09-8 EINECS: 212-464-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...