Cadmium(II)iodid, ultra trocken ≥99,9985% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr


Unter Argon in Ampullen verpackt

35683.03 35683.06 35683.14
35683.03EA 39.1 EUR
35683.03 35683.06 35683.14
Cadmium(II)iodid, ultra trocken ≥99,9985% (Metall-Basis)
Cadmium(II)iodid

Purity: 99.9985% CAS No.: 7790-80-9 Molecular Formula: CdI2 Formula Weight: 366.21 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.67 Appearance: White to yellow Odour: Odourless Melting Point: 387-389° Boiling Point: 787° Merck Reference: 14,1621 UN No.: 2570 MDL No.: MFCD00010920

Formel: CdI₂
Siedepunkt: 787 °C
Schmelzpunkt: 387…389 °C
Dichte: 5,67
Lagertemperatur: Raumtemperatur
MDL: MFCD00010920
CAS-Nummer: 7790-80-9
EINECS: 232-223-6
UN: 2570
ADR: 6.1,III
Merck Index: 14,1621

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR