Chrom(III)chlorid, wasserfrei ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chromium trichloride

35691.06 35691.14 35691.22
35691.06EA 40.5 EUR
35691.06 35691.14 35691.22
Chrom(III)chlorid, wasserfrei ≥99,9% (Metall-Basis)
Chrom(III)chlorid

Purity: 99.9% CAS No.: 10025-73-7 Molecular Formula: CrCl3 Formula Weight: 158.35 Physical Form: Flake Density: 2.87 Appearance: Violet Melting Point: ca 1150° Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00010948

Formel: Cl₃Cr
Molecular Weight: 158.35 g/mol
Schmelzpunkt: ∼ 1150 °C
Dichte: 2.87
Lagertemperatur: Raumtemperatur
MDL: MFCD00010948
CAS-Nummer: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 14,2222

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR