Gallium(II)chlorid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Unter Argon in Ampullen verpackt
35696.03
35696.03EA
101
EUR
35696.03
Gallium(II)chlorid, ultra trocken ≥99,999% (Metall-Basis)
Gallium(II)chlorid
Purity: 99.999%
CAS No.: 24597-12-4
Molecular Formula: Ga2Cl4
Formula Weight: 281.26
Physical Form: Powder, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Appearance: White
Odour: Acidic
Melting Point: 164°
Boiling Point: 535°
UN No.: 3260
MDL No.: MFCD00151526
Formel:
Ga₂Cl₄ Molecular Weight: 281,26 g/mol Siedepunkt: 535 °C Schmelzpunkt: 164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151526 CAS-Nummer: 13498-12-9 EINECS: 246-338-4 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...