Blei(II)bromid, dry_ultra 99,999 Metals basis
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Bleidibromid
ampuled under argon
35703.14
35703.14EA
868
EUR
35703.14
Blei(II)bromid, dry_ultra 99,999 Metals basis
Blei(II)bromid
Purity: 99.999%
CAS No.: 10031-22-8
Molecular Formula: Br2Pb
Formula Weight: 367.01
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 6.66
Appearance: White
Odour: Odourless
Melting Point: 373°
Boiling Point: 916°
Merck Reference: 14,5403
UN No.: 2291
MDL No.: MFCD00011156
Formel:
Br₂Pb Molecular Weight: 367.01 g/mol Siedepunkt: 916 °C Schmelzpunkt: 373 °C Dichte: 6.66 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011156 CAS-Nummer: 10031-22-8 EINECS: 233-084-4 UN: 2291 ADR: 6.1 III Merck Index: 14,5403 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...