Praseodymtribromid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Unter Argon in Ampullen verpackt
35714.04
35714.09
35714.04EA
215
EUR
35714.04
35714.09
Praseodymtribromid, ultra trocken ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Praseodymtribromid
Purity: 99.99%
CAS No.: 13536-53-3
Molecular Formula: PrBr3
Formula Weight: 380.63
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00049762
Formel:
Br₃Pr Schmelzpunkt: 691 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049762 CAS-Nummer: 13536-53-3 EINECS: 236-893-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...