Natriumchlorid, ultra trocken ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Saline, NaCl, Sea salt, Halite
Unter Argon in Ampullen verpackt
35716.14
35716.22
35716.06
35716.14EA
145
EUR
35716.14
35716.22
35716.06
Natriumchlorid, ultra trocken ≥99,99% (Metall-Basis)
Natriumchlorid
Purity: 99.99%
CAS No.: 7647-14-5
Molecular Formula: NaCl
Formula Weight: 58.44
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 2.165
Refractive Index: 1.5442
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
pH: 6-9
Melting Point: 801°
Boiling Point: 1413°
Merck Reference: 14,8599
MDL No.: MFCD00003477
Formel:
NaCl Molecular Weight: 58.44 g/mol Siedepunkt: 1413 °C Schmelzpunkt: 801 Dichte: 2.165 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003477 CAS-Nummer: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,8599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...