Thallium(I)iodid, ultra trocken ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Thalliumiodid

Unter Argon in Ampullen verpackt

35722.09 35722.18 35722.30
35722.09EA 100 EUR
35722.09 35722.18 35722.30
Thallium(I)iodid, ultra trocken ≥99,999% (Metall-Basis)
Thallium(I)iodid

Purity: 99.999% CAS No.: 7790-30-9 Molecular Formula: TlI Formula Weight: 331.27 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 7.1 Appearance: Yellow Odour: Odourless Melting Point: 440° Boiling Point: 824° Merck Reference: 14,9264 UN No.: 1707 MDL No.: MFCD00011279

Formel: ITl
Molecular Weight: 331.29 g/mol
Siedepunkt: 824 °C
Schmelzpunkt: 440
Dichte: 7.1
Lagertemperatur: Raumtemperatur
MDL: MFCD00011279
CAS-Nummer: 7790-30-9
EINECS: 232-199-7
UN: 1707
ADR: 6.1 II
Merck Index: 14,9264

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR