Titan(IV)iodid, ultra trocken ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Titantetraiodid
Unter Argon in Ampullen verpackt
35724.04
35724.04EA
154
EUR
35724.04
Titan(IV)iodid, ultra trocken ≥99,99% (Metall-Basis)
Titan(IV)iodid
Purity: 99.99%
CAS No.: 7720-83-4
Molecular Formula: TiI4
Formula Weight: 555.52
Physical Form: Powder, Ampouled under argon
Density: 4.30
Melting Point: 150°
UN No.: 3260
MDL No.: MFCD00043077
Formel:
I₄Ti Molecular Weight: 555.5 g/mol Siedepunkt: 360 Schmelzpunkt: 150 Dichte: 4.30 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043077 CAS-Nummer: 7720-83-4 EINECS: 231-754-0 UN: 3260 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...