Zinkchlorid, ultra trocken ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chlorzink, Zinkdichlorid, Zink(II)chlorid

Unter Argon in Ampullen verpackt

35726.06 35726.14 35726.22
35726.06EA 87.8 EUR
35726.06 35726.14 35726.22
Zinkchlorid, ultra trocken ≥99,999% (Metall-Basis)
Zinkchlorid

Purity: 99.999% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 Physical Form: Powder, Ampouled under argon Density: 2.91 Melting Point: 290° Boiling Point: 732° Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295

Formel: ZnCl₂
Siedepunkt: 732 °C
Schmelzpunkt: 290 °C
Dichte: 2,91
Lagertemperatur: Raumtemperatur
MDL: MFCD00011295
CAS-Nummer: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 14,10132

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR