Cäsiumiodid, dry_ultra 99,998 Metals basis
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cesiumiodide
ampuled under argon
35729.03
35729.09
35729.30
35729.18
35729.03EA
47.7
EUR
35729.03
35729.09
35729.30
35729.18
Cäsiumiodid, dry_ultra 99,998 Metals basis
Cäsiumiodid
Purity: 99.998%
CAS No.: 7789-17-5
Molecular Formula: CsI
Formula Weight: 259.81
Physical Form: -10 Mesh Beads, Ampouled under argon
Refractive Index: 1.7876
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 626°
Boiling Point: 1280°
Merck Reference: 14,2014
MDL No.: MFCD00010962
Formel:
CsI Molecular Weight: 259.81 g/mol Siedepunkt: 1280 °C Schmelzpunkt: 626 °C Dichte: 3.21 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010962 CAS-Nummer: 7789-17-5 EINECS: 232-145-2 Merck Index: 14,2014 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...