Chrom(III)fluorid ≥98% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Chromium trifluoride

35741.04 35741.09
35741.04EA 239 EUR
35741.04 35741.09
Chrom(III)fluorid ≥98% (Metall-Basis)
Chrom(III)fluorid

Purity: 99.98% CAS No.: 7788-97-8 Molecular Formula: CrF3 Formula Weight: 108.99 Physical Form: Powder Density: 3.8 Appearance: Dark green Odour: Odourless Melting Point: 1000° Merck Reference: 14,2223 UN No.: 1756 MDL No.: MFCD00016045

Formel: CrF₃
Schmelzpunkt: >1000 °C
Dichte: 3,8
Lagertemperatur: Raumtemperatur
MDL: MFCD00016045
CAS-Nummer: 7788-97-8
EINECS: 232-137-9
UN: 1756
ADR: 8,II
Merck Index: 14,2223

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR