Natriumborhydrid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

35788.22 35788.36
35788.22EA 77.8 EUR
35788.22 35788.36
Natriumborhydrid ≥97%
Natriumtetrahydroborat

Purity: 97+% CAS No.: 16940-66-2 Molecular Formula: NaBH4 Formula Weight: 37.83 Physical Form: -10+40 Mesh Granules Density: 1.074 Appearance: White Odour: Weak Melting Point: 400° dec. Solubility: Soluble in water, ammonia and amines. Insoluble in ether Merck Reference: 14,8592 UN No.: 1426 MDL No.: MFCD00003518


As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases

Formel: NaBH₄
Schmelzpunkt: 400 °C (Zersetzung)
Dichte: 1,074
Flammpunkt: 69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003518
CAS-Nummer: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3,I
Merck Index: 14,8592

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR