Bismut(III)sulfid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

35834.18 35834.30
35834.18EA 166 EUR
35834.18 35834.30
Bismut(III)sulfid ≥99,9% (Metall-Basis)
Bismut(III)sulfid

Purity: 99.9% CAS No.: 1345-07-9 Molecular Formula: Bi2S3 Formula Weight: 514.15 Physical Form: Chunks/Crystals Density: 7.7 Appearance: Brown/black MDL No.: MFCD00014204

Formel: Bi₂S₃
Schmelzpunkt: 763 °C
Dichte: 7,7
Lagertemperatur: Raumtemperatur
MDL: MFCD00014204
CAS-Nummer: 1345-07-9
EINECS: 215-716-0
Merck Index: 12,01329

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR