Cerdisilicid ≥99,9%

Lieferant: Thermo Fisher Scientific

Synonyme: Cerium silicide

35836.06 35836.14
35836.06EA 95.3 EUR
35836.06 35836.14
Cerdisilicid ≥99,9%
Cerium disilicide

Purity: 99.9% CAS No.: 12014-85-6 Molecular Formula: CeSi2 Formula Weight: 196.29 Physical Form: 3-15mm Pieces Density: 5.31 Appearance: Grey Odour: Odourless Melting Point: 1620° MDL No.: MFCD00168082

Formel: CeSi₂
Molecular Weight: 196.29 g/mol
Dichte: 5.31
Lagertemperatur: Raumtemperatur
MDL: MFCD00168082
CAS-Nummer: 12014-85-6
EINECS: 234-602-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR