Titan ≥99,7% (Metall-Basis), Folie, geglüht, Dicke 0.032 mm (0.0013 in)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
35881.WF
35881.WG
35881.WH
35881.WFEA
110
EUR
35881.WF
35881.WG
35881.WH
Titan ≥99,7% (Metall-Basis), Folie, geglüht, Dicke 0.032 mm (0.0013 in)
Titanium
Purity: 99.7%
CAS No.: 7440-32-6
Physical Form: ≈8.3g/23x25cm
Density: 4.50
Odour: Odourless
Melting Point: 1660°
Boiling Point: 3287°
MDL No.: MFCD00011264
Formel:
Ti Molecular Weight: 47.88 g/mol Siedepunkt: 3287 °C Schmelzpunkt: 1660 Dichte: 4.50 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011264 CAS-Nummer: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...