Crucibles, tantalum
Lieferant: Thermo Fisher Scientific
35892.KT
35892.KTEA 0 EUR
35892.KT
Crucibles, tantalum
Tiegel
These crucibles are manufactured from tantalum. Available in low form and straight wall types.
  • Melting point of 2996 °C
  • Superior strength at high temperatures
  • Excellent corrosion resistance
Tantalum exhibitis a resistance to acid attack comparable to that of glass and platinum.