Crucibles for automatic fusion equipment, zirconium
Lieferant: Thermo Fisher Scientific
36020.KT
36020.KTEA 280 EUR
36020.KT
Crucibles for automatic fusion equipment, zirconium
Tiegel
These crucibles are manufactured from zirconium. Available with slide-on ring, snap-on ring or flanged rim.
  • Slide-on ring: A removable ring that slides up, fitting approximately 5,08 mm below the top of the crucible
  • Snap-on ring: A removable ring that snap-fits into a machined groove located approximately 5,08 mm below the top of the crucible
  • Flanged rim: A formed rim, integral to the crucible (not welded on), at the crucible's top edge