Bis(cyclopentadienyl)hafniumdichlorid

Lieferant: Thermo Scientific
Gefahr

Synonyme: Hafnocene dichloride

unter Argon abgefüllt

36105.06
36105.06EA 220 EUR
36105.06
Bis(cyclopentadienyl)hafniumdichlorid
Bis(cyclopentadienyl)hafniumdichlorid

CAS No.: 12116-66-4 Molecular Formula: C10H10Cl2Hf Formula Weight: 379.59 Physical Form: Crystalline, Packaged under argon Sensitivity: Air and Moisture Sensitive Melting Point: 230-233° UN No.: 3261 MDL No.: MFCD00001438

Formel: C₁₀H₁₀Cl₂Hf
Molecular Weight: 379,58 g/mol
Schmelzpunkt: 230…233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001438
CAS-Nummer: 12116-66-4
EINECS: 235-177-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR