Gold ≥99,95% (Metall-Basis), Folie, Dicke 0.5 mm (0.02 in)
Lieferant: Thermo Fisher Scientific
Synonyme:
Aurum
36148.FF
36148.FI
36148.FFEA
1615
EUR
36148.FF
36148.FI
Gold ≥99,95% (Metall-Basis), Folie, Dicke 0.5 mm (0.02 in)
Gold
Purity: 99.95%
CAS No.: 7440-57-5
Physical Form: ≈6g/25x25mm
Odour: Odourless
MDL No.: MFCD00003436
Formel:
Au Molecular Weight: 196.97 g/mol Siedepunkt: 2800 Schmelzpunkt: 1063 Dichte: 19.3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003436 CAS-Nummer: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...