2-(3-Aminophenylsulphonyl)ethanol hydrochloride 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ100

361526-1G
361526-1GEA 209 EUR
361526-1G
2-(3-Aminophenylsulphonyl)ethanol hydrochloride 97%, Sigma-Aldrich®
2-(3-Aminophenylsulphonyl)ethanol hydrochloride
Formel: H₂NC₆H₄SO₂CH₂CH₂OH·HCl
Molecular Weight: 237.70 g/mol
Schmelzpunkt: 210 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00134194
CAS-Nummer: 19076-03-0

Bestellen

Spezifikation Testergebnisse

Appearance White to brown powder, crystals or crystalline powder
Infrared spectrum Conforms to structure
Titration 96.5 - 103.5% HClO4
High pressure liquid 96.5% (minimum)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR