ω,4-Dibromacetophenon, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Gefahr
|
Synonyme:
2-Brom-1-(4-bromphenyl)-1-ethanon, Tau Protein Kinase I Inhibitor, 2,4'-Dibromacetophenon, Glycogen Synthase Kinase-3β Inhibitor VII, GSK-3β Inhibitor VII, 4-Bromphenacylbromid, TPK I Inhibitor, α,4-Dibromacetophenon
361548-5
361548-5EA
221
EUR
361548-5
ω,4-Dibromacetophenon, Millipore®
ω,4-Dibromacetophenon
Formel:
C₈H₆Br₂O Molecular Weight: 277.94 g/mol Siedepunkt: 310 Schmelzpunkt: 107…108 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00000200 CAS-Nummer: 99-73-0 EINECS: 202-783-6 UN: 3261 ADR: 8 II Merck Index: 13,01412 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...