(2'Z,3'E)-6-Bromoindirubin-3'-oxime, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: GSK 3 Inhibitor IX, BIO, 6BIO, 6-Bromoindirubin-3'-oxime, (2Z,3E)-6'-Bromo-3-(hydroxyimino)-[2,3'-biindolinylidene]-2'-one

361550-1 361550-10MG 361550-30MG
361550-1EA 221 EUR
361550-1 361550-10MG 361550-30MG
(2'Z,3'E)-6-Bromoindirubin-3'-oxime, Millipore®
(2'Z,3'E)-6-Bromoindirubin-3'-oxime

A cell-permeable bis-indolo (indirubin) compound that acts as a highly potent, selective, reversible and ATP-competitive inhibitor of GSK-3α/β (IC50 = 5 nM).


Inhibition of GSK by BIO has been shown to result in the activation of Wnt-signaling pathway and sustained pluripotency in human and murine ESCs (embryonic stem cells). Reported to maintain self-renewal in human and mouse embryonic stem cells.

Formel: C₁₆H₁₀BrN₃O₂
Molecular Weight: 356.18 g/mol
MDL: MFCD08705318
CAS-Nummer: 667463-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR