Dimethylpolysiloxane set of analytical standards, molecular weight series
Lieferant: Merck
361879-1SET
361879-1SETEA 368 EUR
361879-1SET
Dimethylpolysiloxane set of analytical standards, molecular weight series
Standards Gaschromatographie-Standards
Clear, colorless fluid polymer useful as a stationary phase in gas chromatography and as an anti-foaming agent.
It was used in a study performed to understand its molecular weight and molecular weight distribution by off-line coupling of GPC with MALDI mass spectrometer.
Kit components
Mw 40000
Mw 100000
Mw 270000
Standards: Exact values can be taken from the accompanying certificate of analysis.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f21ae9001b503855ae4dcd70d240836869bf2bb7&vid=f21ae9001b503855ae4dcd70d240836869bf2bb7&prid=web&clid=SAPDAM