Tridemorph, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

36199-100MG
36199-100MGEA 57.7 EUR
36199-100MG
Tridemorph, Supelco®
Tridemorph
Formel: C₁₉H₃₉NO
CAS-Nummer: 24602-86-6
EINECS: 246-347-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0950c361ad4d54933b9939e5f7ebb7f94f9d0e24&vid=d020e642a9a9671e16c7850ff6ceb7ea8850818f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR