Titan(IV)oxid anatas ≥99,6% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pigment White 6, Titansäureanhydrid anatas, Anatas, Titandioxid anatas
36199.14
36199.22
36199.36
36199.14EA
44.3
EUR
36199.14
36199.22
36199.36
Titan(IV)oxid anatas ≥99,6% (Metall-Basis)
Titan(IV)oxid anatas
Purity: 99.6%
CAS No.: 1317-70-0
Molecular Formula: O2Ti
Formula Weight: 79.90
Physical Form: -325 Mesh Powder
Density: 3.9
Merck Reference: 14,9472
MDL No.: MFCD00011269
Formel:
TiO₂ Molecular Weight: 79.88 g/mol Dichte: 3.9 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011269 CAS-Nummer: 1317-70-0 EINECS: 215-280-1 Merck Index: 14,9472 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...