di-Ammoniumoxalat Monohydrat 99,0-101,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.0-101.0%
CAS No.: 6009-70-7
Molecular Formula: (COONH4)2.H2O
Formula Weight: 142.11 (124.10anhy)
Physical Form: Granular
Density: 1.50
Appearance: White
Melting Point: 70° dec.
Solubility: Soluble in water. Slightly soluble in alcohol
Beilstein Reference: 3759830
Merck Reference: 14,537
UN No.: 3288
MDL No.: MFCD00149694
In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals
Formel:
(NH₄)₂C₂O₄·H₂O Siedepunkt: 70 °C (1013 hPa) Schmelzpunkt: 70 °C (Zersetzung) Dichte: 1,50 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149694 CAS-Nummer: 6009-70-7 EINECS: 214-202-3 UN: 3288 ADR: 6.1,III Merck Index: 14,537 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...