Ammoniumcarbonat ≥30% (NH₃-Basis) ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ammoniumcarbonat, Hirschhornsalz

36229.36 36229.A3
36229.36EA 53 EUR
36229.36 36229.A3
Ammoniumcarbonat ≥30% (NH₃-Basis) ACS
Ammoniumcarbonat (enthält Ammoniumcarbamat)

CAS No.: 506-87-6 Molecular Formula: Approx. (NH4)2CO3 Formula Weight: 96.09 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Ammonia-like Melting Point: 60° subl. Solubility: Slowly soluble in water (1:4 portion). Decomposes in hot water Beilstein Reference: 3627235 Merck Reference: 14,508 MDL No.: MFCD00010890


In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds

Formel: (NH₄)₂CO₃
Molecular Weight: 96.09 g/mol
Schmelzpunkt: 60 °C (Subl.)
Dichte: 1.5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010890
CAS-Nummer: 506-87-6
EINECS: 208-058-0
Merck Index: 14,508

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR